Learn More
Medchemexpress LLC Amelparib hydrochloride | 98.4% | 379.88 | 10 MG
SDP
Catalog No. 5000436803
Encompass_Preferred
Amelparib hydrochloride is a potent, orally active, and water-soluble inhibitor of PARP-1. It inhibits PARP-1 activity and cellular PAR formation, acting as a potential neuroprotective agent.
  • Potent, orally active, and water-soluble inhibitor of PARP-1
  • Inhibits PARP-1 activity (IC50 = 18.5 nM)
  • Inhibits cellular PAR formation (IC50 = 10.7 nM)
  • Potential neuroprotective agent
  • Potential for the research of acute ischaemic stroke
Catalog No. 50-004-36803 Supplier Medchemexpress LLC Supplier No. HY116218C10MG
Amelparib hydrochloride is a potent, orally active, and water-soluble inhibitor of PARP-1. It inhibits PARP-1 activity and cellular PAR formation, acting as a potential neuroprotective agent.
  • Potent, orally active, and water-soluble inhibitor of PARP-1
  • Inhibits PARP-1 activity (IC50 = 18.5 nM)
  • Inhibits cellular PAR formation (IC50 = 10.7 nM)
  • Potential neuroprotective agent
  • Potential for the research of acute ischaemic stroke