Learn More
Medchemexpress LLC Amelparib hydrochloride | 98.4% | 379.88 | C19H26ClN3O3 | 5 MG
SDP
Catalog No. 5000264499
Encompass_Preferred
Amelparib hydrochloride is the hydrochloride salt form of a PARP-1 inhibitor used as a research reagent to investigate DNA damage response and PARP-related pathways. It is supplied as a high-purity solid suitable for biochemical and cellular assays.
  • Potent PARP-1 inhibitor activity
  • Hydrochloride salt for improved water solubility
  • High purity (98.4%)
  • Molecular weight 379.88 g/mol
  • Suitable for biochemical and cellular research applications
  • Supplied as a 5 mg solid package
Catalog No. 50-002-64499 Supplier Medchemexpress LLC Supplier No. HY116218C5MG
Amelparib hydrochloride is the hydrochloride salt form of a PARP-1 inhibitor used as a research reagent to investigate DNA damage response and PARP-related pathways. It is supplied as a high-purity solid suitable for biochemical and cellular assays.
  • Potent PARP-1 inhibitor activity
  • Hydrochloride salt for improved water solubility
  • High purity (98.4%)
  • Molecular weight 379.88 g/mol
  • Suitable for biochemical and cellular research applications
  • Supplied as a 5 mg solid package