Recombinant human amelogenin X isoform supplied as an N-terminal SUMO fusion with a 6xHis affinity tag, expressed in E. coli and purified to >90% by reducing SDS-PAGE. The protein corresponds to amino acids M17-D191 (≈35.9 kDa) and is provided in small research quantities for in vitro studies of enamel biomineralization, protein interaction assays, and structural or biochemical analyses. Datasheet, SDS, and certificate of analysis are available.
- Recombinant protein corresponding to amino acids M17-D191.
- N-terminal SUMO and 6xHis tags to facilitate purification and detection.
- Expressed in E. coli for non-glycosylated protein production.
- Purity greater than 90% by reducing SDS-PAGE.
- Approximate molecular weight of 35.9 kDa.
- Available in small microgram sizes suitable for biochemical assays.
- Datasheet, SDS, and certificate of analysis available for quality control.