Learn More
Medchemexpress LLC HY-14809 5mg Medchemexpress, Amenamevir CAS:841301-32-4 Purity:>98%
SDP
Catalog No. 502081644
Encompass_Preferred
Medchemexpress, HY-14809 5mg Amenamevir CAS:841301-32-4 Amenamevir is a helicase-primase inhibitor which has potent antiviral activity against HSVs with an EC50 of 14 ng/mL. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1644 Supplier Medchemexpress LLC Supplier No. HY148095MG
Medchemexpress, HY-14809 5mg Amenamevir CAS:841301-32-4 Amenamevir is a helicase-primase inhibitor which has potent antiviral activity against HSVs with an EC50 of 14 ng/mL. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.