Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Amentoflavone 7,4',7'',4'''-tetramethyl ether | 3778-25-4 | 1 MG
SDP

Catalog No. 5000420864
Encompass_Preferred

Amentoflavone 7,4',7'',4'''-tetramethyl ether is a biflavone, a natural product that can be isolated from Selaginella moellendorffii. It is intended for research use only.

  • Molecular weight: 594.56
  • Formula: C34H26O10
  • Appearance: Solid
  • Color: Off-white to yellow
  • Purity: 97.0%
  • Structure classification: Flavonoids, biflavones
  • Initial source: Plants (Selaginella moellendorffii Hieron.)
  • Natural product with potential selective cytotoxicity

Catalog No. 50-004-20864 Supplier Medchemexpress LLC Supplier No. HYN27561MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Amentoflavone 7,4',7'',4'''-tetramethyl ether is a biflavone, a natural product that can be isolated from Selaginella moellendorffii. It is intended for research use only.

  • Molecular weight: 594.56
  • Formula: C34H26O10
  • Appearance: Solid
  • Color: Off-white to yellow
  • Purity: 97.0%
  • Structure classification: Flavonoids, biflavones
  • Initial source: Plants (Selaginella moellendorffii Hieron.)
  • Natural product with potential selective cytotoxicity

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.