Learn More
Grainger American Beauty Tools Soldering Iron Tip Cleaner, Brass, Large, 1 ea
Catalog No. 502795842
Encompass_Preferred
This brass sponge tip cleaner is designed for efficient removal of contaminants from soldering iron tips. It allows for effective tip maintenance without the need for water, helping to preserve tip life and performance.
  • Cleans soldering iron tips without water
  • Includes a stand for stable operation
  • Constructed from durable brass
  • Compatible with specific soldering iron stations
Catalog No. 50-279-5842 Supplier Grainger Supplier No. 19YP76
This brass sponge tip cleaner is designed for efficient removal of contaminants from soldering iron tips. It allows for effective tip maintenance without the need for water, helping to preserve tip life and performance.
  • Cleans soldering iron tips without water
  • Includes a stand for stable operation
  • Constructed from durable brass
  • Compatible with specific soldering iron stations