Learn More
Grainger American Standard Infection Control Wall-Hung Sink, Vitreous China, 20 in. x 17 in., 1 ea
Catalog No. 502765933
Encompass_Preferred
The infection control wall-hung sink is designed to minimize splashing and maintain a hygienic environment in demanding healthcare and commercial settings. Crafted from durable vitreous china, this wall-mounted sink features a deep basin with sloped ledges and a rear-offset drain to guide water away from the drain grid. The integrated antimicrobial surface inhibits the growth of stain- and odor-causing bacteria, mold, and mildew, while integral side and back splashes provide additional protection against contamination.
  • Deep basin and sloped ledges minimize splashing
  • Antimicrobial surface protects against bacteria, mold, and mildew
  • Offset, non-pooling drain with sealed overflow
  • Includes P-trap and offset drain with antimicrobial coating
  • Removable acrylic shroud with mounting hardware included
  • Lead-law compliant and approved for commercial use
Catalog No. 50-276-5933 Supplier Grainger Supplier No. 16U008
The infection control wall-hung sink is designed to minimize splashing and maintain a hygienic environment in demanding healthcare and commercial settings. Crafted from durable vitreous china, this wall-mounted sink features a deep basin with sloped ledges and a rear-offset drain to guide water away from the drain grid. The integrated antimicrobial surface inhibits the growth of stain- and odor-causing bacteria, mold, and mildew, while integral side and back splashes provide additional protection against contamination.
  • Deep basin and sloped ledges minimize splashing
  • Antimicrobial surface protects against bacteria, mold, and mildew
  • Offset, non-pooling drain with sealed overflow
  • Includes P-trap and offset drain with antimicrobial coating
  • Removable acrylic shroud with mounting hardware included
  • Lead-law compliant and approved for commercial use