Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amerex™ 10 lb. (4.5kg) Multipurpose Extinguisher

Catalog No. 19029145 Shop All Amerex Corporation Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Model:
B441
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
19-029145 B441
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-029145 Supplier Amerex™ Supplier No. 14966
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Wall-mount bracket.

Contains specially fluidized and siliconized monoammonium-phosphate-based dry chemical

Choice of aluminum or chrome-plated brass valves.

  • For Class A, B or C fires
  • Contain specially fluidized and siliconized monoammonium-phosphate-based dry chemical
  • Choice of aluminum or chrome-plated brass valves

Certifications

UL rating: 4-A:80-B:C. FM approved.

Specifications

Product Type Multi-purpose Fire Extinguisher
Material Steel
Capacity (English) 10 lb.
Capacity (Metric) 4.5 kg
Dimensions (L x W x H) 5 x 8.75 x 20.5 in. (12.7 x 22.2 x 52.1 cm)
Discharge Time 20 sec.
Height (English) 20.5 in.
Height (Metric) 52.1 cm
Includes Wall mount bracket
Length (English) 5 in.
Length (Metric) 12.7 cm
Model B441
Range 15 to 21 ft.
Reagent Type ABC dry chemical
Temperature Range (English) -65° to +120°F
Temperature Range (Metric) -53.8°C to +48.8°C
UL Rating 4A:80B:C
Width (English) 8.75 in.
Width (Metric) 22.2 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.