Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amerex™ Class D Extinguishers

Catalog No. 19029185 Shop All Amerex Corporation Products
Encompass_Preferred
Change view
Click to view available options
Discharge Time:
24 sec.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Discharge Time
19-029185 24 sec.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-029185 Supplier Amerex™ Supplier No. 16882
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For combustible metals fires

Both extinguishers feature dependable drawn steel cylinders, all-metal valve construction and special corrosion resistant yellow (Class D) color-coded paint finish.

Sodium Chloride Extinguisher

  • Contains a specially blended sodium chloride-based dry powder extinguishing agent
  • Heat from fire causes sodium chloride to cake and form a crust, excluding air and dissipating heat from burning metal
  • Successfully extinguishers fires involving magnesium, sodium, potassium and sodium-potassium alloys
  • Zirconium, uranium, titanium, and powdered aluminum fires can be controlled and extinguished

Copper Extinguisher

  • Contains a copper extinguishing agent specially developed by the U.S. Navy for fighting lithium and lithium alloy fires
  • Copper compound smothers the fire and provides an excellent heat sink for dissipating heat
  • Copper powderis superior to other known fire extinguishing agents for lithium

Order Info

Capacity: 30 lb. (13.6kg). Shipping weight: 53 lb. (24kg). Dimensions: 7D x 16W x 27.75H in. (17.8 x 40.6 x 70.5cm). Larger capacity and wheeled extinguishers also available. Contact your Fisher Safety Customer Service Representative for ordering information.

Certifications

FM approved.

Warranty and Services

Six years.

Specifications

Product Type Class D Fire Extinguisher
Material Steel
Dimensions (L x W x H) 8 x 13 x 35 in. (20.3 x 33.02 x 88.9 cm)
Discharge Time 24 sec.
Height (English) 35 in.
Height (Metric) 88.9 cm
Includes Wall bracket
Length (English) 8 in.
Length (Metric) 20.3 cm
Model B570
Range 3 to 6 ft.
Reagent Type Sodium Chloride
Temperature Range (English) -40° to +120°F
Temperature Range (Metric) -40°C to +48.8°C
Weight (English) 30 lb.
Weight (Metric) 13.6 kg
Width (English) 13 in.
Width (Metric) 33.02 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.