Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amerex™ CO2 Fire Extinguishers

Catalog No. p-39199 Shop All Amerex Corporation Products
Encompass
Change view
Click to view available options
Discharge Time:
10 sec.
12.5 sec.
Model:
322
331
UL Rating:
10B:C
5B:C
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Discharge Time Model UL Rating
19-029091 12.5 sec. 331 10B:C
19-860-537 10 sec. 322 5B:C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-029091 Supplier Amerex™ Supplier No. 01703
Only null left
Add to Cart
Add to Cart

Includes

Wall-mount bracket, hose and horn.

Electrically nonconductive agent effective against Class B flammable liquid fires

  • Clean, noncontaminating, odorless
  • Safe for use on clothing, equipment, valuable documents or food
  • Lightweight aluminum cylinder
  • All-metal valve construction
  • Barcoded and bilingual labels
  • Model 19-806-120 for use in MRI facilities

5 lb. CO2 Extinguisher

  • Capacity: 5 lb. (2.3kg)
  • Shipping weight: 14 lb. (6.3kg)
  • Dimensions: 5.25D x 8.25W x 17.75 in.H (13.3 x 20.9 x 45.1cm)

10 lb. CO2 Extinguisher

  • Capacity: 10 lb. (4.5kg)
  • Shipping weight: 28.75 lb. (12.8kg)

15 lb. CO2 Extinguisher

  • Capacity: 15 lb. (6.8kg)
  • Shipping weight: 37.75 lb. (12.8kg)

Certifications

UL listed. FM approved. USCG approved with bracket listed on UL label.

Specifications

Product Type Fire Extinguisher
Material Aluminum
Dimensions (L x W x H) 7 x 12 x 30 in. (17.7 x 30.48 x 76.2 cm)
Discharge Time 12.5 sec.
Height (English) 30 in.
Height (Metric) 76.2 cm
Includes Wall mount bracket, hose and horn
Length (English) 7 in.
Length (Metric) 17.7 cm
Model 331
Range 3 to 8 ft.
Reagent Type Carbon Dioxide
Temperature Range (English) -22° to +120°F
Temperature Range (Metric) -30°C to +48.8°C
UL Rating 10B:C
Weight (English) 15 lb.
Weight (Metric) 6.8 kg
Width (English) 12 in.
Width (Metric) 30.48 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.