Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amerex™ Purple K Dry Chemical Fire Extinguishers

Catalog No. p-3202529 Shop All Amerex Corporation Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Discharge Time:
28 sec.
Capacity (English):
20 lb.
Material:
Steel, Anodized Aluminum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Discharge Time Capacity (English) Material
19-860-507 28 sec. 20 lb. Steel, Anodized Aluminum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-860-507 Supplier Amerex™ Supplier No. 11215
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Fire-fighting agent for the oil, gas, chemical and utilities industries

  • Contain specially fluidized and siliconized potassium bicarbonate dry chemical
  • Particularly effective on Class B flammable liquids and pressurized gases
  • Barcoded and bilingual labels
  • Wall-mount drawn steel cylinders and chrome-plated brass valve construction
  • Temperature ranges from -65° to 120°F

Certifications

FM approved. UL rated. USCG approved (with bracket)

Warranty and Services

6 years

Specifications

Product Type Fire Extinguisher
Material Steel, Anodized Aluminum
Capacity (English) 20 lb.
Capacity (Metric) 9.07 kg
Dimensions (L x W x H) 7 x 10.25 x 24 in. (17.7 x 26 x 60.9 cm)
Discharge Time 28 sec.
Height (English) 24 in.
Height (Metric) 60.9 cm
Length (English) 7 in.
Length (Metric) 17.7 cm
Model A413
Range 15 to 21 ft.
Reagent Type Purple K
Temperature Range (English) -65° to +120°F
Temperature Range (Metric) -53.8°C to +48.8°C
UL Rating 120B:C
Width (English) 10.25 in.
Width (Metric) 26 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.