Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amerex™ Water Fire Extinguisher

Catalog No. 19029081 Shop All Amerex Corporation Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Capacity (English):
2.5 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
19-029081 2.5 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-029081 Supplier Amerex™ Supplier No. 00100
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Product Type Fire Extinguisher
Material Stainless Steel
Capacity (English) 2.5 gal.
Capacity (Metric) 9.4 L
Dimensions (L x W x H) 7 x 9 x 24.5 in. (17.7 x 22.86 x 62.2 cm)
Discharge Time 55 sec.
Height (English) 24.5 in.
Height (Metric) 62.2 cm
Includes Wall bracket
Length (English) 7 in.
Length (Metric) 17.7 cm
Model B416
Range 45 to 55 ft.
Reagent Type Water
Temperature Range (English) 40° to 120°F
Temperature Range (Metric) 4.4°C to 48.8°C
UL Rating 2A
Width (English) 9 in.
Width (Metric) 22.86 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.