Learn More
American Biotech Supply AMERICAN BIOTECH SUPPLY Undercounter Refrigerator, 4.6 Cu. Ft., Built-In, ADA Compliant
Catalog No. NC3985941
Encompass_Preferred
This 4.6 Cu. Ft. undercounter refrigerator is designed for built-in applications with front breathing ventilation and is ADA compliant (under 32 inches height). It features a microprocessor controller for precise temperature management and uses eco-friendly R600a hydrocarbon refrigerants, meeting EPA/SNAP, UL, and ASHRAE guidelines.
  • Microprocessor temperature control with digital display and remote alarms.
  • Audible/visual high/low temperature alarms with air and glycol bottle probes.
  • Adjustable operating temperature range: 1°C to 10°C.
  • Three shelves (two adjustable), keyed door lock, and magnetic door gasket.
  • Heavy-duty handle, leveling legs, HFC-free foam insulation, and forced draft.
  • UL/C-UL Listed, ADA Compliant, and Energy Star Certified.
Catalog No. NC3985941 Supplier American Biotech Supply Supplier No. ABTHCUCBI0404ADA
This 4.6 Cu. Ft. undercounter refrigerator is designed for built-in applications with front breathing ventilation and is ADA compliant (under 32 inches height). It features a microprocessor controller for precise temperature management and uses eco-friendly R600a hydrocarbon refrigerants, meeting EPA/SNAP, UL, and ASHRAE guidelines.
  • Microprocessor temperature control with digital display and remote alarms.
  • Audible/visual high/low temperature alarms with air and glycol bottle probes.
  • Adjustable operating temperature range: 1°C to 10°C.
  • Three shelves (two adjustable), keyed door lock, and magnetic door gasket.
  • Heavy-duty handle, leveling legs, HFC-free foam insulation, and forced draft.
  • UL/C-UL Listed, ADA Compliant, and Energy Star Certified.