Learn More
GSC Go Science Crazy Friction Rods and Silk Pad
SCI_ED
Catalog No. S43222 Shop All GSC Products
Click to view available options
Product Type:
Friction Rod
Catalog No. Product Type
S43222 Friction Rod
1 options
Catalog No. S43222 Supplier GSC Go Science Crazy Supplier No. 603110
Learn about electrostatics.
All the essential materials for exploration in electrostatics.
  • Topics include static electricity, polarity and friction studies
  • Use a variety of friction rods, friction pads and pith balls
  • Combination of materials determines charge distribution after contact
Specifications
Material Glass
For Use With (Application) Static electricity, polarity and friction studies
Product Type Friction Rod
Length (English) 12 in.
Length (Metric) 30.48 cm
WARNING: Cancer - www.P65Warnings.ca.gov