Learn More
American Fiber and Finishing Purewipe™ Disposable Cheesecloth Wipers
SDP
Catalog No. 0666517
Click to view available options
Length (English):
10 yd.
Catalog No. Length (English)
06-665-17 10 yd.
1 options
This item is not returnable. View return policy
Catalog No. 06-665-17 Supplier American Fiber and Finishing Supplier No. 590200
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
This item is not returnable. View return policy
100% woven cotton
  • Ideal for all-purpose cleaning. Grade 10: open weave, 20 × 12 threads/sq. in.
  • Biodegradable and incinerable
  • Silicone-free
  • No ozone-depleting agents
Order Info
In polyethylene bags. Pack of 10 yd. (9m).
Specifications
Product Type PureWipe™ Cheesecloth Wipes
Length (English) 10 yd.
Material Woven Cotton
Disposable Single-use
For Use With (Application) For all cleaning purpose