Learn More
American Fiber and Finishing Purewipe™ Heavy-duty Cheesecloth Wipers
SDP
Catalog No. 0666525B
Click to view available options
Length (English):
70 yd.
Catalog No. Length (English)
06-665-25B 70 yd.
1 options
This item is not returnable. View return policy
Catalog No. 06-665-25B Supplier American Fiber and Finishing Supplier No. 588039/DUPL
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
This item is not returnable. View return policy
Ideal as a press cloth for crystals in oil analysis
Useful for cleaning, polishing, straining, filtering waxes. Heavy-duty dust cloth. Washable. Grade 50: closely knit weave, 28 × 24 threads/sq. in.
  • 100% woven cotton
  • Biodegradable and incinerable
  • Silicone-free
  • No ozone-depleting agents
Order Info
In handifold bolt of 70 yd. (64m).
Specifications
Product Type PureWipe™ Heavy Duty Cheesecloth Wipes
Length (English) 70 yd.
Material Woven Cotton
For Use With (Application) Cleaning, polishing, straining, filtering waxes, ideal as a press cloth for crystals in oil analysis