Learn More
American Fiber and Finishing Purewipe™ Individual Cheesecloth Wipers
SDP
Catalog No. 0666525A
Click to view available options
Length (English):
1 yd.
Catalog No. Length (English)
06-665-25A 1 yd.
1 options
This item is not returnable. View return policy
Catalog No. 06-665-25A Supplier American Fiber and Finishing Supplier No. 588108
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
This item is not returnable. View return policy
Useful for filtering waxes
Use as press cloth for crystals in oil analysis or as a dust cloth. Washable. Grade 40: fine weave, 24 × 20 threads/sq. in.
  • 100% cotton
  • Biodegradable and incinerable
  • Silicone-free
  • No ozone-depleting agents
Order Info
In convenient polyethylene bags. Pack of two 1 yd. (91cm) cloths.
Specifications
Product Type PureWipe™ Washable Cheesecloth Wipes
Length (English) 1 yd.
Material Woven Cotton
For Use With (Application) Filtering wax, press cloth for crystals in oil analysis or as a dust cloth