Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

American Innotek Brief Relief™ Field Lavatory System

Catalog No. 19805954 Shop All American Innotek Inc. Products
Change view
Click to view available options
Color:
Gray
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-805-954 Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-805-954 Supplier American Innotek Supplier No. PQ2000

missing translation for 'validMainframeMsg'

Includes

  • Water-resistant, durable, lightweight privacy tent
  • Commode container with soft, folding seat
  • 10-pack of Brief Relief Daily Restroom Kits
  • Each Daily Restroom Kit contains 2 urine pouches and 1 solid waste bag

Portable, single-stall field toilet and shelter

Perfect for work crews on the go—system can be set up virtually anywhere.

  • Requires little storage space and only two minutes of set-up time
  • Waste disposal pouches are trash container safe, spill-proof, leak-proof, and odorless, eliminating the need for costly dumping and servicing fees

Order Info

Unlimited shelf life

Specifications

Product Type Field Lavatory System
Color Gray
For Use With (Application) Perfect for work crews on the go system can be set up virtually anywhere
Includes Privacy tent, commode container, folding seat, 10 pack of brief relief daily restroom kits
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.