Learn More
American Innotek Brief Relief™ Pouch
Catalog No. 19805952 Shop All American Innotek Inc. Products
Change view
Click to view available options
Product Type:
Pouch
Catalog No. Product Type
19-805-952 Pouch
1 options
This item is not returnable. View return policy
Catalog No. 19-805-952 Supplier American Innotek Supplier No. BR608
This item is not returnable. View return policy
For use by both men and women
  • Pocket size, single use, sealable bag for liquid human waste
  • Absorbs liquid waste and odor until fully contained in a gelled state
  • Waste is approved for landfill disposal
  • Includes a sanitary wipe for easy cleanup after use
Specifications
Product Type Pouch
For Use With (Application) For use by both men and women
Includes Sanitary wipe for easy cleanup after use
Size Pocket