Learn More
Med Vet International Ameroid Constrictor 7.5mm, J0659E, Each
Catalog No. 5000584606
Encompass_Preferred
For the surgical treatment of porto caval or hepatic shunts. This specially designed product is used to encircle a blood vessel. A special material (hygroscopic compressed casein) inside the donut shaped constrictor imbibes fluid over time. It slowly occludes the blood vessel. A small key is used to lock it in place after it is slipped over the blood vessel. Sizing: about a 25% reduction in the blood vessel diameter should be achieved upon initial surgical placement for the best results. Sterile. Diameter of inner circle: 7.5mm
Catalog No. 50-005-84606 Supplier Med Vet International Supplier No. JOR659E
For the surgical treatment of porto caval or hepatic shunts. This specially designed product is used to encircle a blood vessel. A special material (hygroscopic compressed casein) inside the donut shaped constrictor imbibes fluid over time. It slowly occludes the blood vessel. A small key is used to lock it in place after it is slipped over the blood vessel. Sizing: about a 25% reduction in the blood vessel diameter should be achieved upon initial surgical placement for the best results. Sterile. Diameter of inner circle: 7.5mm