Learn More
Med Vet International Ameroid Constrictors 6.5mm, J0659C
Catalog No. 5000585268
Encompass_Preferred
For the surgical treatment of porto caval or hepatic shunts.This specially designed product is used to encircle a blood vessel. A special material (hygroscopic compressed casein) inside the donut shaped constrictor imbibes fluid over time. It slowly occludes the blood vessel.A small key is used to lock it in place after it is slipped over the blood vessel.Sizing: about a 25% reduction in the blood vessel diameter should be achieved upon initial surgical placement for the best results. Sterile.Diameter of inner circle: 6.5mm
Catalog No. 50-005-85268 Supplier Med Vet International Supplier No. JOR659C
For the surgical treatment of porto caval or hepatic shunts.This specially designed product is used to encircle a blood vessel. A special material (hygroscopic compressed casein) inside the donut shaped constrictor imbibes fluid over time. It slowly occludes the blood vessel.A small key is used to lock it in place after it is slipped over the blood vessel.Sizing: about a 25% reduction in the blood vessel diameter should be achieved upon initial surgical placement for the best results. Sterile.Diameter of inner circle: 6.5mm