Learn More
Med Vet International Ameroid Constrictors 6mm, J0659B
Catalog No. 5000584454
Encompass_Preferred
For the surgical treatment of porto caval or hepatic shunts.This specially designed product is used to encircle a blood vessel. A special material (hygroscopic compressed casein) inside the donut shaped constrictor imbibes fluid over time. It slowly occludes the blood vessel.A small key is used to lock it in place after it is slipped over the blood vessel.Sizing: about a 25% reduction in the blood vessel diameter should be achieved upon initial surgical placement for the best results. Sterile.Diameter of inner circle: 6.0mm
Catalog No. 50-005-84454 Supplier Med Vet International Supplier No. JOR659B
Add to Cart
Add to Cart
For the surgical treatment of porto caval or hepatic shunts.This specially designed product is used to encircle a blood vessel. A special material (hygroscopic compressed casein) inside the donut shaped constrictor imbibes fluid over time. It slowly occludes the blood vessel.A small key is used to lock it in place after it is slipped over the blood vessel.Sizing: about a 25% reduction in the blood vessel diameter should be achieved upon initial surgical placement for the best results. Sterile.Diameter of inner circle: 6.0mm