Learn More
Cayman Chemical amethyl FentnylhydroCLOr 100ug
SDP
Catalog No. 502918764
Encompass_Preferred
An analytical reference standard categorized as an opioid; approximately as potent as fentanyl and 57 times more potent than morphine for its analgesic properties in mice; provided as a DEA exempt preparation; intended for research and forensic applications
Catalog No. 50-291-8764 Supplier Cayman Chemical Supplier No. 38654100 UG
An analytical reference standard categorized as an opioid; approximately as potent as fentanyl and 57 times more potent than morphine for its analgesic properties in mice; provided as a DEA exempt preparation; intended for research and forensic applications