Learn More
Medchemexpress LLC Ametryn | 834-12-8 | MFCD00055398 | 98.6% | 250 MG
SDP
Catalog No. 5000407065
Encompass_Preferred
Ametryn, a member of the Triazine chemical family, is a herbicide which inhibits photosynthesis and other enzymatic processes. It is effective against annual broadleaf weeds and grasses.
  • Belongs to the Triazine chemical family
  • Functions as a herbicide
  • Inhibits photosynthesis
  • Inhibits enzymatic processes
  • Effective against annual broadleaf weeds
  • Effective against grasses
Catalog No. 50-004-07065 Supplier Medchemexpress LLC Supplier No. 250MGHYB0866
Ametryn, a member of the Triazine chemical family, is a herbicide which inhibits photosynthesis and other enzymatic processes. It is effective against annual broadleaf weeds and grasses.
  • Belongs to the Triazine chemical family
  • Functions as a herbicide
  • Inhibits photosynthesis
  • Inhibits enzymatic processes
  • Effective against annual broadleaf weeds
  • Effective against grasses