Learn More
Selleck Chemical LLC Amezinium
SDP
Catalog No. 5000004582
Encompass_Preferred
Amezinium is a sympathomimetic used for its vasopressor effects in the treatment of hypotensive states It is a MAO inhibitor antagonizes the response to tyramine and blocks neuronal uptake of noradrenaline
Catalog No. 50-000-04582 Supplier Selleck Chemical LLC Supplier No. S646225MG
Amezinium is a sympathomimetic used for its vasopressor effects in the treatment of hypotensive states It is a MAO inhibitor antagonizes the response to tyramine and blocks neuronal uptake of noradrenaline