Learn More
Medchemexpress LLC Amfenac-d5 (sodium) | 61941-56-8 | 98.90% | 282.28 | 5 MG
SDP
Catalog No. 5000387058
Encompass_Preferred
Amfenac-d5 sodium is deuterated-labeled Amfenac sodium, an orally active and potent anti-inflammatory compound possessing antipyretic and analgesic properties. It can be used for the research of acute and chronic inflammation. This compound can also be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Deuterated-labeled Amfenac sodium
  • Orally active
  • Potent anti-inflammatory compound
  • Possesses antipyretic and analgesic properties
  • Can be used for the research of acute and chronic inflammation
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-87058 Supplier Medchemexpress LLC Supplier No. HY17479BS5MG
Amfenac-d5 sodium is deuterated-labeled Amfenac sodium, an orally active and potent anti-inflammatory compound possessing antipyretic and analgesic properties. It can be used for the research of acute and chronic inflammation. This compound can also be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Deuterated-labeled Amfenac sodium
  • Orally active
  • Potent anti-inflammatory compound
  • Possesses antipyretic and analgesic properties
  • Can be used for the research of acute and chronic inflammation
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS