Learn More
Selleck Chemical LLC Amfenac Sodium Monohydrate 50mg 61618-27-7
SDP
Supplier:  Selleck Chemical LLC S414950MG
Encompass_Preferred
Amfenac Sodium monohydrate is a non-steroidal analgesic anti-inflammatory drug with acetic acid moiety. The IC50 values for COX1 and COX2 is 250 nM and 150 nM, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8373