Learn More
Medchemexpress LLC Amfenac (Sodium Hydrate) | 61618-27-7 | 98.3% | 295.27 | 1 ML
SDP
Catalog No. 5000386788
Encompass_Preferred
Amfenac Sodium Hydrate is a COX-2 inhibitor. It decreases the proliferation rate of cells transfected to express COX-2 and inhibits nitric oxide production by macrophages. It also inhibits the release of B-glucuronidase and aggregation of polymorphonuclear leukocytes (PMNs).
  • Decreases the proliferation rate of cells transfected to express COX-2.
  • Partially overcomes the inhibition of nitric oxide production by macrophages caused by melanoma conditioned medium.
  • Inhibits the release of β-glucuronidase.
  • Causes inhibition of aggregation of polymorphonuclear leukocytes (PMNs).
Catalog No. 50-003-86788 Supplier Medchemexpress LLC Supplier No. HY17479A10MM/1ML
Amfenac Sodium Hydrate is a COX-2 inhibitor. It decreases the proliferation rate of cells transfected to express COX-2 and inhibits nitric oxide production by macrophages. It also inhibits the release of B-glucuronidase and aggregation of polymorphonuclear leukocytes (PMNs).
  • Decreases the proliferation rate of cells transfected to express COX-2.
  • Partially overcomes the inhibition of nitric oxide production by macrophages caused by melanoma conditioned medium.
  • Inhibits the release of β-glucuronidase.
  • Causes inhibition of aggregation of polymorphonuclear leukocytes (PMNs).