Learn More
Apexbio Technology LLC Amfenac Sodium Monohydrate 61618-27-7 10mM (in 1mL DMSO)
SDP
Catalog No. 5000091269
Encompass_Preferred
Amfenac Sodium Monohydrate (CAS 61618-27-7) is a synthetic non-steroidal anti-inflammatory compound structurally characterized by an acetic acid moiety It functions by inhibiting cyclooxygenase (COX) enzymes consequently suppressing prostaglandin synthesis and mitigating inflammatory processes and pain signaling Amfenac Sodium Monohydrate is utilized in biomedical research to investigate inflammatory pathways and to evaluate non-steroidal analgesic drug effects in cellular and animal models of inflammation Its well-defined inhibitory mechanism makes it a valuable tool for elucidating COX-mediated physiological and pathological responses
Catalog No. 50-000-91269 Supplier Apexbio Technology LLC Supplier No. B16565.1
Amfenac Sodium Monohydrate (CAS 61618-27-7) is a synthetic non-steroidal anti-inflammatory compound structurally characterized by an acetic acid moiety It functions by inhibiting cyclooxygenase (COX) enzymes consequently suppressing prostaglandin synthesis and mitigating inflammatory processes and pain signaling Amfenac Sodium Monohydrate is utilized in biomedical research to investigate inflammatory pathways and to evaluate non-steroidal analgesic drug effects in cellular and animal models of inflammation Its well-defined inhibitory mechanism makes it a valuable tool for elucidating COX-mediated physiological and pathological responses