Learn More
Selleck Chemical LLC Amfenac Sodium Monohydrate S4149-1g
SDP
Catalog No. 503109115
Encompass_Preferred
Amfenac Sodium monohydrate is a non-steroidal analgesic anti-inflammatory drug with acetic acid moiety The IC50 values for COX1 and COX2 is 250 nM and 150 nM respectively
Catalog No. 50-310-9115 Supplier Selleck Chemical LLC Supplier No. S41491G
Amfenac Sodium monohydrate is a non-steroidal analgesic anti-inflammatory drug with acetic acid moiety The IC50 values for COX1 and COX2 is 250 nM and 150 nM respectively