Learn More
Cayman Chemical Amfenac (sodium salt hydrate), Purity: ≥98%, CAS: 61618-27-7, 1 g
SDP
Catalog No. 502992507
Encompass_Preferred
A COX inhibitor (IC50s = 15.3 and 20.4 nM for the recombinant human COX-1 and COX-2, respectively) and the active metabolite of nepafenac; reduces the migration of 92.1 uveal melanoma cells at 150 nM, as well as proliferation of the same cells when used in combination with ranibizumab
Catalog No. 50-299-2507 Supplier Cayman Chemical Supplier No. 306271 G
A COX inhibitor (IC50s = 15.3 and 20.4 nM for the recombinant human COX-1 and COX-2, respectively) and the active metabolite of nepafenac; reduces the migration of 92.1 uveal melanoma cells at 150 nM, as well as proliferation of the same cells when used in combination with ranibizumab