Learn More
Aviva Systems Biology Corp AMFR Antibody - C-terminal region (ARP42995_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42995_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against AMFR. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human AMFR. Alternate names: GP78, RNF45ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58386