Learn More
Medchemexpress LLC HY-101565 1mg Medchemexpress, AMG-176 CAS:1883727-34-1 Purity:>98%
SDP
Catalog No. 501932170
Encompass_Preferred
Medchemexpress, HY-101565 1mg AMG-176 CAS:1883727-34-1 AMG-176 is a potent, selective and orally bioavailable MCL-1 inhibitor, with a K i of 0.13 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2170 Supplier Medchemexpress LLC Supplier No. HY1015651MG
Medchemexpress, HY-101565 1mg AMG-176 CAS:1883727-34-1 AMG-176 is a potent, selective and orally bioavailable MCL-1 inhibitor, with a K i of 0.13 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.