Learn More
Selleck Chemical LLC AMG-208 10mg 1002304-34-8
SDP
Catalog No. 508571
Encompass_Preferred
AMG 208 is a highly selective dual c-Met and RON inhibitor with IC50 of 9 nM for c-Met. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-857-1 Supplier Selleck Chemical LLC Supplier No. S131610MG
AMG 208 is a highly selective dual c-Met and RON inhibitor with IC50 of 9 nM for c-Met. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.