Learn More
Medchemexpress LLC AMG 333 | 1416799-28-4 | 99.8% | 453.32 | 1 ML
SDP
Catalog No. 5000438543
Encompass_Preferred
AMG 333 is an orally active and highly selective TRPM8 antagonist with an IC50 of 13 nM. It is for research use only, not for sale to patients.
  • Orally active and highly selective TRPM8 antagonist
  • IC50 of 13 nM (TRPM8)
  • Inhibits wet-dog shake behavior in Icilin-induced Sprague-Dawley rat model
  • Blocks blood pressure elevation in cold stimulation-induced Sprague-Dawley rat model
  • Effectively blocks the TRPM8 target
  • Demonstrates significant dose-dependent inhibition of wet-dog shake behavior
  • Shows significant inhibition of cold-induced blood pressure elevation
  • Potential clinical candidate for the treatment of migraine
Catalog No. 50-004-38543 Supplier Medchemexpress LLC Supplier No. HY11270310MM/1ML
AMG 333 is an orally active and highly selective TRPM8 antagonist with an IC50 of 13 nM. It is for research use only, not for sale to patients.
  • Orally active and highly selective TRPM8 antagonist
  • IC50 of 13 nM (TRPM8)
  • Inhibits wet-dog shake behavior in Icilin-induced Sprague-Dawley rat model
  • Blocks blood pressure elevation in cold stimulation-induced Sprague-Dawley rat model
  • Effectively blocks the TRPM8 target
  • Demonstrates significant dose-dependent inhibition of wet-dog shake behavior
  • Shows significant inhibition of cold-induced blood pressure elevation
  • Potential clinical candidate for the treatment of migraine