Learn More
Selleck Chemical LLC AMG-333
SDP
Catalog No. 5000002082
Encompass_Preferred
AMG-333 is a potent and highly selective antagonist of TRPM8 with IC50 values of 13 nM and 30 nM for human TRPM8 and rat TRPM8
Catalog No. 50-000-02082 Supplier Selleck Chemical LLC Supplier No. S66385MG
AMG-333 is a potent and highly selective antagonist of TRPM8 with IC50 values of 13 nM and 30 nM for human TRPM8 and rat TRPM8