Learn More
Medchemexpress LLC Amg-337 | 1173699-31-4 | 99.4% | 463.46 | 200 MG
SDP
Catalog No. 5000388108
Encompass_Preferred
AMG-337 is a potent, orally active, and selective MET kinase inhibitor. It effectively inhibits the phosphorylation of MET and its downstream effectors in MET-amplified cancer cell lines, leading to the inhibition of MET-dependent cell proliferation and the induction of apoptosis. This compound has demonstrated activity in both in vitro and in vivo studies, including various tumor xenograft models.
  • Potent and selective MET kinase inhibitor
  • Inhibits MET-dependent cell proliferation
  • Induces apoptosis in cancer cell lines
  • Effective in various tumor xenograft models
Catalog No. 50-003-88108 Supplier Medchemexpress LLC Supplier No. HY18696200MG
AMG-337 is a potent, orally active, and selective MET kinase inhibitor. It effectively inhibits the phosphorylation of MET and its downstream effectors in MET-amplified cancer cell lines, leading to the inhibition of MET-dependent cell proliferation and the induction of apoptosis. This compound has demonstrated activity in both in vitro and in vivo studies, including various tumor xenograft models.
  • Potent and selective MET kinase inhibitor
  • Inhibits MET-dependent cell proliferation
  • Induces apoptosis in cancer cell lines
  • Effective in various tumor xenograft models