Learn More
Selleck Chemical LLC AMG-458 S2747-50mg
SDP
Catalog No. 503111802
Encompass_Preferred
AMG 458 is a potent c-Met inhibitor with Ki of 1 2 nM 350-fold selectivity for c-Met than VEGFR2 in cells
Catalog No. 50-311-1802 Supplier Selleck Chemical LLC Supplier No. S274750MG
AMG 458 is a potent c-Met inhibitor with Ki of 1 2 nM 350-fold selectivity for c-Met than VEGFR2 in cells