Learn More
TARGETMOL CHEMICALS INC AMG-458 5MG
SDP
Catalog No. 502579404
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AMG-458 is a potent c-Met inhibitor with Ki of 1.2 nM ~350-fold selectivity for c-Met than VEGFR2 in cells. purity: 99%
Catalog No. 50-257-9404 Supplier TARGETMOL CHEMICALS INC Supplier No. T63785MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AMG-458 is a potent c-Met inhibitor with Ki of 1.2 nM ~350-fold selectivity for c-Met than VEGFR2 in cells. purity: 99%