Learn More
Selleck Chemical LLC AMG-458 S2747-5mg
SDP
Catalog No. 502173099
Encompass_Preferred
AMG 458 is a potent c-Met inhibitor with Ki of 1 2 nM 350-fold selectivity for c-Met than VEGFR2 in cells
Catalog No. 50-217-3099 Supplier Selleck Chemical LLC Supplier No. S27475MG
AMG 458 is a potent c-Met inhibitor with Ki of 1 2 nM 350-fold selectivity for c-Met than VEGFR2 in cells