Learn More
Selleck Chemical LLC AMG 487
SDP
Supplier:  Selleck Chemical LLC S868225MG
Encompass_Preferred
AMG 487 is an orally active and selective CXC chemokine receptor 3 (CXCR3) antagonist that inhibits the binding of IP-10 (CXCL10) and ITAC (CXCL11) to CXCR3 with IC50 of 8 0 nM and 8 2 nM respectively
Catalog No. 50-000-01204