Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC AMG 487 | 473719-41-4 | C32H28F3N5O4 | 1 ML
SDP

Catalog No. 5000372697
Encompass_Preferred

AMG 487 is an orally active and selective antagonist of CXC chemokine receptor 3 (CXCR3), inhibiting the binding of CXCL10 and CXCL11 to CXCR3 with IC50s of 8.0 and 8.2 nM, respectively. It is intended for research use only.

  • Orally active and selective CXCR3 antagonist
  • Inhibits CXCL10 and CXCL11 binding to CXCR3
  • Purity of 99.89%
  • Molecular weight of 603.59
  • White to yellow solid appearance
  • Inhibits CXCR3-mediated cell migration
  • Inhibits calcium mobilization

Catalog No. 50-003-72697 Supplier Medchemexpress LLC Supplier No. HY1531910MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AMG 487 is an orally active and selective antagonist of CXC chemokine receptor 3 (CXCR3), inhibiting the binding of CXCL10 and CXCL11 to CXCR3 with IC50s of 8.0 and 8.2 nM, respectively. It is intended for research use only.

  • Orally active and selective CXCR3 antagonist
  • Inhibits CXCL10 and CXCL11 binding to CXCR3
  • Purity of 99.89%
  • Molecular weight of 603.59
  • White to yellow solid appearance
  • Inhibits CXCR3-mediated cell migration
  • Inhibits calcium mobilization

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.