Learn More
Selleck Chemical LLC AMG 487 5mg
SDP
Supplier:  Selleck Chemical LLC S86825MG
Encompass_Preferred
AMG 487 is an orally active and selective CXC chemokine receptor 3 (CXCR3) antagonist that inhibits the binding of IP-10 (CXCL10) and ITAC (CXCL11) to CXCR3 with IC50 of 8.0 nM and 8.2 nM, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3381