Learn More
TARGETMOL CHEMICALS INC AMG 487 5MG
SDP
Catalog No. 502880758
Encompass_Preferred
Also available in 1 mL, 1 mg, 2 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. AMG 487 is a potent and selective antagonist of chemokine (C-X-C motif) receptor 3 (CXCR3) which inhibits the binding of CXCL10 and CXCL11 to CXCR3 with IC50s of 8.0 and 8.2 nM, respectively. Purity 99.89%
Catalog No. 50-288-0758 Supplier TARGETMOL CHEMICALS INC Supplier No. T10297L5MG
Also available in 1 mL, 1 mg, 2 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. AMG 487 is a potent and selective antagonist of chemokine (C-X-C motif) receptor 3 (CXCR3) which inhibits the binding of CXCL10 and CXCL11 to CXCR3 with IC50s of 8.0 and 8.2 nM, respectively. Purity 99.89%