Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC AMG 487 (S-enantiomer) | 473720-30-8 | C32H28F3N5O4 | 25 MG
SDP

Catalog No. 5000375418
Encompass_Preferred

AMG 487 S-enantiomer is the S enantiomer of AMG 487. It functions as an antagonist of the chemokine receptor CXCR3, targeting the GPCR/G Protein and Immunology/Inflammation pathways.

  • Purity: 99.94%
  • Appearance: Off-white to light yellow solid
  • Storage (powder): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 2 years, -20°C for 1 year
  • In vitro solubility: 100 mg/mL in DMSO

Catalog No. 50-003-75418 Supplier Medchemexpress LLC Supplier No. 25MGHY15319B
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AMG 487 S-enantiomer is the S enantiomer of AMG 487. It functions as an antagonist of the chemokine receptor CXCR3, targeting the GPCR/G Protein and Immunology/Inflammation pathways.

  • Purity: 99.94%
  • Appearance: Off-white to light yellow solid
  • Storage (powder): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 2 years, -20°C for 1 year
  • In vitro solubility: 100 mg/mL in DMSO

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.