Learn More
Medchemexpress LLC AMG-510 10MG 99.6%
SDP
Catalog No. NC1769907
Encompass_Preferred
AMG-510 10mg 99.6% -20°C, stored under nitrogen* In solvent : -80°C, 6 months; -20°C, 1 month (stored under nitrogen)
Catalog No. NC1769907 Supplier Medchemexpress LLC Supplier No. HY11427710MG
AMG-510 10mg 99.6% -20°C, stored under nitrogen* In solvent : -80°C, 6 months; -20°C, 1 month (stored under nitrogen)