Learn More
Medchemexpress LLC HY-114277 5mg Medchemexpress, AMG-510 CAS:2296729-00-3 Purity:>98%
SDP
Catalog No. 502028834
Encompass_Preferred
Medchemexpress, HY-114277 5mg AMG-510 CAS:2296729-00-3 AMG-510 is a first-in-class, orally bioavailable, and selective KRAS G12C covalent inhibitor. AMG-510 irreversibly inhibits KRAS G12C by locking it in an inactive GDP-bound state. AMG-510 is the first KRAS G12C inhibitor in clinical development and leads to the regression of KRAS G12C tumors[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8834 Supplier Medchemexpress LLC Supplier No. HY1142775MG
Medchemexpress, HY-114277 5mg AMG-510 CAS:2296729-00-3 AMG-510 is a first-in-class, orally bioavailable, and selective KRAS G12C covalent inhibitor. AMG-510 irreversibly inhibits KRAS G12C by locking it in an inactive GDP-bound state. AMG-510 is the first KRAS G12C inhibitor in clinical development and leads to the regression of KRAS G12C tumors[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.