Learn More
Medchemexpress LLC HY-13440 5mg Medchemexpress, AMG 511 CAS:1253573-53-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY134405MG
Encompass_Preferred
Medchemexpress, HY-13440 5mg AMG 511 CAS:1253573-53-3 AMG 511 is a potent and orally available pan inhibitor of class I PI3Ks, with Kis of 4 nM, 6 nM, 2 nM and 1 nM for PI3Kα, β, δ and γ, respectively. AMG 511 significantly suppresses PI3K signaling that is indicated by p-Akt (Ser473) decrease. AMG 511 exhibits anti-tumor activity in mouse glioblastoma xenograft model. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0312