Learn More
TARGETMOL CHEMICALS INC AMG 517 25MG
SDP
Catalog No. 502579402
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AMG 517 is an effective and specific TRPV1 antagonist antagonizes proton (IC50 0.76 nM) capsaicin (IC50 0.62 nM) and heat activation (IC50 1.3 nM) of TRPV1. purity: 99%
Catalog No. 50-257-9402 Supplier TARGETMOL CHEMICALS INC Supplier No. T637925MG
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AMG 517 is an effective and specific TRPV1 antagonist antagonizes proton (IC50 0.76 nM) capsaicin (IC50 0.62 nM) and heat activation (IC50 1.3 nM) of TRPV1. purity: 99%