Learn More
Medchemexpress LLC HY-10634 5mg Medchemexpress, AMG 517 CAS:659730-32-2 Purity:>98%
SDP
Catalog No. 501968337
Encompass_Preferred
Medchemexpress, HY-10634 5mg AMG 517 CAS:659730-32-2 AMG 517 is a potent and selective vanilloid receptor-1 (TRPV1) antagonist with an IC50 of 0.5 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8337 Supplier Medchemexpress LLC Supplier No. HY106345MG
Medchemexpress, HY-10634 5mg AMG 517 CAS:659730-32-2 AMG 517 is a potent and selective vanilloid receptor-1 (TRPV1) antagonist with an IC50 of 0.5 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.